Masterflex® Fitting, Polypropylene, Straight, Hose Barb Reducer, 1/4" ID x 5/32" ID; 10/PK
Catalog # MFLX40622-26
Supplier: Avantor Fluid Handling
CE certificate
About this item
Best for low-pressure applications.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Temperture range 9 to 180 °F (-13 to +66 °C)
These economical straight barbed reducing fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.
- バーブサイズ/チューブ内径 (ポート1):½"
- バーブサイズ/チューブ内径 (ポート2):⁵⁄₃₂"
- 接続タイプ:Barbed Reducer
- Flow pattern:Straight
- 説明:Fitting, polypropylene, straight, hose barb reducer, int.Ø ½×⁵⁄₃₂";10/pack
- Seq:27
- Environmentally Preferable:
- Model:40622-26
- Sterilisation:Ethylene oxide (ETO) or gamma
- Pk:10
- Fitting material:Polypropylene
Frequently Bought Together