酸化還元(ORP)標準
400 mV (at 25°C), ±5 mV, HDPE bottle, 500 mL
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Avantor offers a wide range of REDOX (ORP) standards, which are produced from high quality reagents.These reference materials can be used as control standards to check the functionality of both the elextrodes and the measuring instruments. The values are temperature dependent, so it is important to control the environment of the measurement. A table of temperatue and corresponding Redox value is available on both the label and the batch specific certificate.
- Accuracy ±5 mV
- Quoted values tested at 25 °C
- Temperature vs Redox value as a table on the bottle label
- Stability excellent throughout shelf life
ORP standards where all values quoted are potentials of platinum electrode v Ag/AgCl reference electrode (3M KCl)
Certificates
Search by Lot NumberSpecifications
Specifications
- パックタイプ:HDPE bottle
- Pk:500 mL
- 説明:400 mV (at 25°C)
- Accuracy:±5 mV



