Order Entry
Japan
Orders LinkContactUsLinkComponent
酸化還元(ORP)標準

酸化還元(ORP)標準

358 mV (at 25°C), ±5 mV, HDPE bottle, 500 mL
Catalog # 91228.270
Supplier:   VWR Chemicals
酸化還元(ORP)標準
Catalog # 91228.270
Supplier:  VWR Chemicals
CAS Number:  
酸化還元(ORP)標準
酸化還元(ORP)標準

358 mV (at 25°C), ±5 mV, HDPE bottle, 500 mL

CAS Number:  
Price
$216.21
Log in for availability
1 * 500 mL
Product Details & Documents

Avantor offers a wide range of REDOX (ORP) standards, which are produced from high quality reagents.These reference materials can be used as control standards to check the functionality of both the elextrodes and the measuring instruments. The values are temperature dependent, so it is important to control the environment of the measurement. A table of temperatue and corresponding Redox value is available on both the label and the batch specific certificate.

  • Accuracy ±5 mV
  • Quoted values tested at 25 °C
  • Temperature vs Redox value as a table on the bottle label
  • Stability excellent throughout shelf life

ORP standards where all values quoted are potentials of platinum electrode v Ag/AgCl reference electrode (3M KCl)

Documents

Specifications

Specifications

  • パックタイプ:
    HDPE bottle
  • Pk:
    500 mL
  • 説明:
    358 mV (at 25°C)
  • Accuracy:
    ±5 mV

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.