Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, シンチレーションバイアル

VWR®, シンチレーションバイアル

20,00 ml, 28×61 mm, Clear, No, 22-400
Catalog # 215-4491
Supplier:   VWR Collection
VWR®, シンチレーションバイアル
Catalog # 215-4491
Supplier:  VWR Collection
VWR®, シンチレーションバイアル
VWR®, シンチレーションバイアル

20,00 ml, 28×61 mm, Clear, No, 22-400

Price
Inquire For Price
Log in for availability
1 * 250 items
Product Details & Documents

VWR® scintillation vials are produced from borosilicate 5.1 expansion glass. They are compatible with all scintillation counters.

  • High resistance against chemicals and thermal shock
  • Low background counts and high ultraviolet transmission
  • Delivered in convenient pack sizes

VWR® scintillation vials are ideal for use in liquid scintillation counting, beta/gamma counting or for sample storage.

Ordering information: Screw caps have to be ordered separately and you can choose the right screw cap for your application from our selection of fitting screw caps with the most common liners, which we offer as accessories.

Packaging: Packed in shrink-wrapped trays in a convenient 250 piece pack size.

Specifications
  • キャップ付き:
    No
  • キャップサイズ:
    22-400
  • Ø×H:
    28×61 mm
  • 容量:
    20,00 ml
  • Colour:
    Clear
  • Seq:
    02
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.