384プレート、J.T.Baker®製
PP plates, Transparent, 5 - 180 µl, -, 200 µl
Catalog # 43004-0200
Supplier: Avantor
Jump to:
Product Details & Documents
Available in PP for automated screening and storage systems.
- Alphanumeric code to assist sample identification
- Square wells for increased volume
- V-bottom for high sample recovery
- High frame (3 mm) for stable and slip-free stacking
J.T.Baker Plate 384 is ideal for small sample volumes.
Documents
Specifications
- Recommended working volume:5 - 180 µl
- Sterile:-
- Colour:Transparent
- 説明:PP plates
- Well volume:200 µl
- Cat. No.:43004-0200
- Environmental certification:ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
- Environmentally Preferable:Low Manufacturing Impact