Orders LinkContactUsLinkComponent
384プレート、J.T.Baker®製
384プレート、J.T.Baker®製
PP plates, Transparent, 5 - 110 µl, -, 120 µl
Catalog # 43004-0120
   Low Manufacturing Impact
Supplier:   Avantor
384プレート、J.T.Baker®製
Catalog # 43004-0120
Supplier:  Avantor
   Low Manufacturing Impact
384プレート、J.T.Baker®製
384プレート、J.T.Baker®製
PP plates, Transparent, 5 - 110 µl, -, 120 µl
Price
$759.61
Log in for availability
1 * 60 items
Product Details & Documents
Available in PP for automated screening and storage systems.
  • Alphanumeric code to assist sample identification
  • Square wells for increased volume
  • V-bottom for high sample recovery
  • High frame (3 mm) for stable and slip-free stacking
J.T.Baker Plate 384 is ideal for small sample volumes.
Specifications
  • Recommended working volume:
    5 - 110 µl
  • Sterile:
    -
  • Colour:
    Transparent
  • 説明:
    PP plates
  • Well volume:
    120 µl
  • Cat. No.:
    43004-0120
  • Environmental certification:
    ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
  • Environmentally Preferable:
    Low Manufacturing Impact