J.T.Baker®プレート96
PS plates, F-bottom, Clear, -, 0,35 ml
: Avantor
J.T.Baker Plate 96 round well plates made from PS are ideal for ELISA applications.
- Alphanumeric coding assists with identification of samples
- Plate shape significantly reduces sample carry-over
- Standardised dimensions (128×85 mm) ensure compatibility with most common laboratory machines
- Sterile:-
- Colour:Clear
- 説明:PS plates, F-bottom
- Well volume:0,35 ml
- Cat. No.:43001-0119
- Environmental certification:ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
- Environmentally Preferable:Low Manufacturing Impact



