J.T.Baker®ロボットチップDX型
300 µl, -, Blue, 1× 4 racks, 108 each
Catalog # 49136-0000
Supplier: Avantor
Jump to:
Product Details & Documents
Suitable for use on Dynex DSX and DS2 instruments. Available in non conductive version, without filter, in container rack, and in CE-IVD marked version for clinical diagnostic applications.
- 108 tips in one optimized container with lid
- Especially developed for the automated sample handling
- Highest quality specifications for the compatibility and cleanliness
- Blue sample tips for visual tip identification
The validation of the tips on the instrument for specific applications is within the responsibility of the user.
Documents
Specifications
- Packed:1× 4 racks, 108 each
- Sterile:-
- 容量:300 µl
- Seq:1
- Tip colour:Blue
- Cat. No.:49136-0000
- Environmental certification:ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
- Environmentally Preferable:Low Manufacturing Impact
Accessories
Product Information
- Catalog #ColourSterileAvailabilityPrice