Orders LinkContactUsLinkComponent
J.T.Baker®ロボットチップDX型
J.T.Baker®ロボットチップDX型
300 µl, -, Blue, 1× 4 racks, 108 each
Catalog # 49136-0000
   Low Manufacturing Impact
Supplier:   Avantor
J.T.Baker®ロボットチップDX型
Catalog # 49136-0000
Supplier:  Avantor
   Low Manufacturing Impact
J.T.Baker®ロボットチップDX型
J.T.Baker®ロボットチップDX型
300 µl, -, Blue, 1× 4 racks, 108 each
Price
$111.26
Log in for availability
1 * 432 items
Product Details & Documents
Suitable for use on Dynex DSX and DS2 instruments. Available in non conductive version, without filter, in container rack, and in CE-IVD marked version for clinical diagnostic applications.
  • 108 tips in one optimized container with lid
  • Especially developed for the automated sample handling
  • Highest quality specifications for the compatibility and cleanliness
  • Blue sample tips for visual tip identification
The validation of the tips on the instrument for specific applications is within the responsibility of the user.
Specifications
  • Packed:
    1× 4 racks, 108 each
  • Sterile:
    -
  • 容量:
    300 µl
  • Seq:
    1
  • Tip colour:
    Blue
  • Cat. No.:
    49136-0000
  • Environmental certification:
    ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
  • Environmentally Preferable:
    Low Manufacturing Impact
Accessories
Product Information
  • Catalog #
    Colour
    Sterile
    Availability
    Price
  • 43001-0031
    Transparent
    -
    Log in for availability
    1 * 250 items - $272.59