マイクロチューブラック、J.T.Baker®、リバーシブルマイクロチューブラック
Individual tubes, 1,2 ml, Clear, PP
45003-0000
: Avantor
Safe sample storage system.
- V bottom for optimal sample recovery
- Individual tubes or strips of eight tubes available
- 96-well capmat and strips of eight plug caps available
- Refillable alphanumeric marked rack
- Non sterile
Microtube racks are supplied with tubes included. Rack can be also filled with individual tubes available separately. They have a footprint in accordance to SBS standard and it is compatible with all current robotic systems.
- 容量:1,2 ml
- Colour:Clear
- 説明:Individual tubes
- Material:PP
- Seq:06