Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, 超純水製造システム、Uシリーズ

VWR®, 超純水製造システム、Uシリーズ

VWR U12 TOC system starter setup (12 L/h)
Catalog # 171-1470
Supplier:   VWR Collection
VWR®, 超純水製造システム、Uシリーズ
Catalog # 171-1470
Supplier:  VWR Collection
VWR®, 超純水製造システム、Uシリーズ
VWR®, 超純水製造システム、Uシリーズ

VWR U12 TOC system starter setup (12 L/h)

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents

This 'two in one' lab water system produces ultrapure (Type I) and RO (Type III) water from tap water. Quality of ultrapure water meets or exceeds ASTM, CLSI, CAP, and ISO Type I water standards.

  • Simple and intuitive to dispense water via the dispenser or central console
  • Central console is an 8 inch waterproof touch screen, providing user with full control and access of system performance
  • One water system can drive several dispensing units ('1 + N mode')
  • Dispensing flow rate is up to 2 L/min and volumetric dispensing is available
  • RFID-tagged consumables for easier tracking of lifetime and performance
  • Wireless communication option to control multiple dispensers and the central console (optional)
  • Real time TOC and resistivity/conductivity monitoring
  • Optional tank circulation mode to maintain stable water quality in the tank
  • Users can see essential information (water quality, system status, warnings) on the dispenser unit and the central console at any time (data stored up to 2 years; downloadable)

The graphic display guides users through system operation, performance, troubleshooting and maintenance, and can be password protected for multiple access levels.

Applications (Type I - ultrapure water): HPLC mobile phase preparation; preparation of reagent blank solutions; sample diluent for GC, HPLC, ICP-MS, AA and other analytical techniques; preparation of buffers and culture media for mammalian cell culture; preparation of molecular biology reagents.

Applications (Type III - RO water): Glassware cleaning, washing machines for glassware, water bath water, autoclave filling, water for laboratory animals.

Certifications: The VWR® water system is delivered with a Certificate of Conformity ensuring that it has been built and tested fully assembled following Standard Operating Procedures, and a Certificate of Calibration for the temperature and resistivity meters built into the system. Consumables are automatically delivered with a Certificate of Quality. Manufacturing site is ISO 9001 (2015) and ISO 140001 certified.

Ordering information: For a complete system, the production unit and mandatory accessories have to be ordered. Other accessories and consumables, including wall mounting brackets, purification cartridges and wireless upgrade kit are available separately.
Please note: For the U12 system you need to order one unit of the RO pack 171-1276 (12 L/h), For the U24 system two units of the RO pack are required 171-1276 (12 L/h). For the U32 system please order two units of the RO pack 171-1277 (16 L/h).

Delivery information: VWR® U12 System starter setup (171-1467) includes 1 × VWR® U12 System, 1 × 0,2 µm PES final filter for VWR®, 1 × VWR® AC Pack , 1 × VWR® P Pack , 1 × VWR® RO Pack, 12 L/H, 1 × VWR® H Pack, 1 × VWR® U Pack, 1 × VWR® Cleaning pack, 1 × Leak protector for VWR®, 1 × Dispenser kit, 1 × Tank vent filter for VWR®, 1 × UV Sanitization module for VWR® tank, and 1 × 30L PE Tank with sensors for VWR®.

VWR® U24 System starter setup (171-1468) includes 1 × VWR® U24 System, 1 × 0,2 µm PES final filter for VWR®, 1 × VWR® AC Pack , 1 × VWR® P Pack , 2 × VWR® RO Pack, 12 L/H, 1 × VWR® H Pack, 1 × VWR® U Pack, 1 × VWR® Cleaning Pack, 1 × Leak protector for VWR®, 1 × Dispenser kit, 1 × Tank vent filter for VWR®, 1 × UV Sanitization module for VWR® tank, and 1 × 30L PE Tank with sensors for VWR®.

VWR® U32 System starter setup (171-1469) includes 1 × VWR® U32 System, 1 × 0,2 µm PES final filter for VWR®, 1 × VWR® AC Pack , 1 × VWR® P Pack , 2 × VWR® RO Pack, 16 L/H, 1 × VWR® H Pack, 1 × VWR® U Pack, 1 × VWR® Cleaning Pack, 1 × Leak protector for VWR®, 1 × Dispenser kit, 1 × Tank vent filter for VWR®, 1 × UV Sanitization module for VWR® tank, and 1 × 30L PE Tank with sensors for VWR®.

VWR® U12 TOC System starter setup (171-1470) includes 1 × VWR® U12 TOC System, 1 × 0,2 µm PES final filter for VWR®, 1 × VWR® AC Pack , 1 × VWR® P Pack , 1 × VWR® RO Pack, 12 L/H, 1 × VWR® H Pack, 1 × VWR® U Pack (Low TOC), 1 × VWR® Cleaning Pack, 1 × Leak protector for VWR®, 1 × Dispenser kit, 1 × Tank vent filter for VWR®, 1 × UV Sanitization module for VWR® tank, and 1 × 30L PE Tank with sensors for VWR®.

VWR® U24 TOC System starter setup (171-1471) includes 1 × VWR® U24 TOC System, 1 × 0,2 µm PES final filter for VWR®, 1 × VWR® AC Pack , 1 × VWR® P Pack , 2 × VWR® RO Pack, 12 L/H, 1 × VWR® H Pack, 1 × VWR® U Pack (Low TOC), 1 × VWR® Cleaning Pack, 1 × Leak protector for VWR®, 1 × Dispenser kit, 1 × Tank vent filter for VWR®, 1 × UV Sanitization module for VWR® tank, and 1 × 30L PE Tank with sensors for VWR®.

VWR® U32 TOC System starter setup (171-1472) includes 1 × VWR® U32 TOC System, 1 × 0,2 µm PES final filter for VWR®, 1 × VWR® AC Pack , 1 × VWR® P Pack , 2 × VWR® RO Pack, 16 L/H, 1 × VWR® H Pack, 1 × VWR® U Pack (Low TOC), 1 × VWR® Cleaning Pack, 1 × Leak protector for VWR®, 1 × Dispenser kit, 1 × Tank vent filter for VWR®, 1 × UV Sanitization module for VWR® tank, and 1 × 30L PE Tank with sensors for VWR®.

Specifications
  • 説明:
    VWR U12 TOC system starter setup (12 L/h)
  • Seq:
    04
  • Production flow rate:
    12 L/h
  • Flow rate:
    Max. 2 l/min
  • Feed water:
    Tap water (conductivity: <2000 µS/cm / TDS: <1000 ppm)
  • DNases:
    <10 pg/ml (with Bio filter)
  • Endotoxin:
    <0,001 EU/ml (with Bio filter)
  • Particles:
    No particles >0,2 μm (with a 0,2 μm final filter or Bio filter)
  • Cat. No.:
    171-1470
  • Microorganisms:
    <0,1 cfu/ml (with a 0,2 μm final filter or Bio filter)
  • Model:
    U12
  • W×D×H:
    320×440×540 mm
  • RNases:
    <0,5 pg/ml (with Bio filter)
  • Resistivity at +25 °C:
    18,2 MΩ·cm (Type I)
  • TOC content:
    <5 ppb (Type I)
Accessories

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1271
    Continuous level sensor, 0 - 2 M, ¼'' G
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1280
    Recirculation pump, 24 VDC
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1282
    UV lamp, 254 nm
    Log in for availability
    1 * 5 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1292
    200 LPH water softener kit dig. contr.
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1375
    Pressure regulator kit
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1476
    Leak protector for VWR®
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1259
    VWR® T pack 
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1270
    Tank UV lamp, 254 nm
    Log in for availability
    1 * 5 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1279
    RO booster pump, 24 VDC
    Log in for availability
    1 * 5 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1288
    LABXT5 EDI module
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1283
    UV lamp, dual wavelength, 185/254 nm
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1289
    LABXT10 EDI module
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1284
    VWR® control console
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1290
    LABXT15 EDI module
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1291
    EDI power module
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1376
    Distribution pump and connection kit
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1377
    Connection kit for Miele, ½" tubing
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1336
    Wall mount kit for dispenser
    Log in for availability
    1 * 1 KIT

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1337
    Wall mount kit for 30L/60 L tank
    Log in for availability
    1 * 1 KIT

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1269
    Tank recirculation upgrade kit with a cartridge
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1287
    Wall mounting kit
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1258
    U pack
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1257
    H pack
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1293
    PreFilter support (no cartridge)
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1294
    Cartridge for Prefilter support
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1276
    RO pack, 12 L/h *
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1277
    RO pack, 16 L/h *
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1281
    Cleaning pack
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1274
    Wireless dispenser upgrade kit
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1275
    Dispenser kit with on-system anchor
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1254
    AC pack
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1255
    P pack
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1256
    P pack for harder water
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1265
    60 L PE tank with sensors
    Log in for availability
    1 * 1 SET

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1263
    Bio filter
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1264
    30 L PE tank with sensors
    Log in for availability
    1 * 1 SET

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1266
    100 L PE tank with continuous level sensor
    Log in for availability
    1 * 1 SET

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1272
    Submersible level sensor, 0 - 3 m
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1273
    Dispenser kit
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1260
    U pack (low TOC)
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1261
    U pack (ICP)
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1286
    Foot switch
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1262
    0,2 μm PES final filter
    Log in for availability
    1 * 1 items

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.