Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, 超純水製造システム、Gシリーズ

VWR®, 超純水製造システム、Gシリーズ

G5 system starter set-up with TOC (5 L/h)
Catalog # 171-1464
Supplier:   VWR Collection
VWR®, 超純水製造システム、Gシリーズ
Catalog # 171-1464
Supplier:  VWR Collection
VWR®, 超純水製造システム、Gシリーズ
VWR®, 超純水製造システム、Gシリーズ

G5 system starter set-up with TOC (5 L/h)

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents

This powerful, intuitive laboratory water purification system provides superior and consistent ultrapure Type I and EDI-based Type II water from tap water. Water quality meets or exceeds ASTM, CLSI, CAP and ISO Type I water standards, respectively.

  • Simple and intuitive to dispense water via the dispenser or central console
  • Central console is an 8 inch waterproof touch screen, providing user with full control and access of system performance
  • One water system can drive several dispensing units ('1 + N mode')
  • Dispensing flow rate is up to 2 L/min and volumetric dispensing is available
  • RFID-tagged consumables for easier tracking of lifetime and performance
  • Wireless communication option to control multiple dispensers and the central console (optional)
  • Real time TOC and resistivity/conductivity monitoring
  • Users can see essential information (water quality, system status, warnings) on the dispenser unit and the central console at any time (data stored up to two years, downloadable)

The graphic display guides users through system operation, performance, troubleshooting and maintenance, and can be password protected for multiple access levels.

Applications (Type I - ultrapure water): HPLC mobile phase preparation; preparation of reagent blank solutions; sample diluent for GC, HPLC, ICP-MS, AA and other analytical techniques; preparation of buffers and culture media for mammalian cell culture; preparation of molecular biology reagents.

Applications (Type II - EDI water): Preparation of chemical and bio-reagents; preparation of culture media; preparation of solutions for chemical analysis (such as HPLC and ICP); clinical analysers; medical devices and equipment rinsing; serum and blood fractionation; ophthalmics.

Certifications: The VWR® water system is delivered with a Certificate of Conformity ensuring that it has been built and tested fully assembled following Standard Operating Procedures, and a Certificate of Calibration for the temperature and resistivity meters built into the system. Consumables are automatically delivered with a Certificate of Quality. Manufacturing site is ISO 9001 (2015) and ISO 140001 certified.

注文情報: For a complete system, the production unit and mandatory accessories have to be ordered. Other accessories and consumables, including wall mounting brackets, purification cartridges and wireless upgrade kit are available separately.
Please note: For the G5 system you need to order one unit of the RO pack 171-1276 (12 L/h), For the G10 system two units of the RO pack are required 171-1276 (12 L/h). For the G15 system please order two units of the RO pack 171-1277 (16 L/h).

納品情報: VWR® G5 system starter set-up (171-1464) includes 1× VWR® G5 system with TOC, 1×0,2 μm PES final filter for VWR®, 1× VWR® AC pack, 1× VWR® P pack, 1× VWR® RO pack, 12 L/H, 1× VWR® U pack (low TOC), 1× VWR® cleaning pack, 1× leak protector for VWR®, 1× dispenser kit, 1× tank vent filter for VWR®, 1× UV sanitisation module for VWR® tank, 1× 30 L PE tank with sensors for VWR®.

VWR® G10 system starter set-up (171-1465) includes 1× VWR® G10 system with TOC, 1×0,2 μm PES final filter for VWR®, 1× VWR® AC pack, 1× VWR® P pack, 2× VWR® RO pack, 12 L/H, 1× VWR® U pack (low TOC), 1× VWR® cleaning pack, 1× leak protector for VWR®, 1× dispenser kit, 1× tank vent filter for VWR®, 1× UV sanitisation module for VWR® tank, 1× 30 L PE tank with sensors for VWR®.

VWR® G15 system starter set-up (171-1466) includes 1× VWR® G15 system with TOC, 1× 0,2 μm PES final filter for VWR®, 1× VWR® AC pack , 1× VWR® P pack, 2× VWR® RO pack, 16 L/H, 1× VWR® U pack (low TOC), 1× VWR® cleaning pack, 1× leak protector for VWR®, 1× dispenser kit, 1× tank vent filter for VWR®, 1× UV sanitisation module for VWR® tank, 1× 30 L PE tank with sensors for VWR®.

Specifications
  • 説明:
    G5 system starter set-up with TOC (5 L/h)
  • Seq:
    01
  • Production flow rate:
    5 L/h
  • Flow rate:
    Max. 2 L/min
  • Feed water:
    Tap water (conductivity: <2000 µS/cm / TDS: <1000 ppm)
  • DNases:
    <10 pg/ml (with Bio filter)
  • Endotoxin:
    <0,001 EU/ml (with Bio filter)
  • Particles:
    No particles >0,2 μm (with a 0,2 μm final filter or Bio filter)
  • Cat. No.:
    171-1464
  • Microorganisms:
    <0,1 cfu/ml (with a 0,2 μm final filter or Bio filter)
  • Model:
    G5
  • W×D×H:
    320×440×540 mm
  • RNases:
    <0,5 pg/ml (with Bio filter)
  • Resistivity at +25 °C:
    18,2 MΩ·cm (Type I)
  • TOC content:
    <5 ppb (Type I)
    <30 ppb (Type II)
Accessories

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1279
    RO booster pump, 24 VDC
    Log in for availability
    1 * 5 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1270
    Tank UV lamp, 254 nm
    Log in for availability
    1 * 5 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1283
    UV lamp, dual wavelength, 185/254 nm
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1290
    LABXT15 EDI module
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1291
    EDI power module
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1292
    200 LPH water softener kit dig. contr.
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1476
    Leak protector for VWR®
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1282
    UV lamp, 254 nm
    Log in for availability
    1 * 5 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1259
    VWR® T pack 
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1284
    VWR® control console
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1288
    LABXT5 EDI module
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1289
    LABXT10 EDI module
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1375
    Pressure regulator kit
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1280
    Recirculation pump, 24 VDC
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1271
    Continuous level sensor, 0 - 2 M, ¼'' G
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1267
    Tank vent filter
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1268
    UV sanitisation module for tank
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1376
    Distribution pump and connection kit
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1377
    Connection kit for Miele, ½" tubing
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1336
    Wall mount kit for dispenser
    Log in for availability
    1 * 1 KIT

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1337
    Wall mount kit for 30L/60 L tank
    Log in for availability
    1 * 1 KIT

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1272
    Submersible level sensor, 0 - 3 m
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1261
    U pack (ICP)
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1286
    Foot switch
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1262
    0,2 μm PES final filter
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1269
    Tank recirculation upgrade kit with a cartridge
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1287
    Wall mounting kit
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1258
    U pack
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1257
    H pack
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1293
    PreFilter support (no cartridge)
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1294
    Cartridge for Prefilter support
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1276
    RO pack, 12 L/h *
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1277
    RO pack, 16 L/h *
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1281
    Cleaning pack
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1274
    Wireless dispenser upgrade kit
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1275
    Dispenser kit with on-system anchor
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1254
    AC pack
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1255
    P pack
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1256
    P pack for harder water
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1265
    60 L PE tank with sensors
    Log in for availability
    1 * 1 SET

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1263
    Bio filter
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1264
    30 L PE tank with sensors
    Log in for availability
    1 * 1 SET

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1266
    100 L PE tank with continuous level sensor
    Log in for availability
    1 * 1 SET

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1273
    Dispenser kit
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 171-1260
    U pack (low TOC)
    Log in for availability
    1 * 1 items

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.