VWR®, 研削ジャー
35 ml, Zirconium oxide, 10 mm, 15 ml
Catalog # 412-0316
Supplier: VWR Collection
Jump to:
Product Details & Documents
These grinding jars can be used with the VWR® mixer mill, beater.
Specifications
- 容量:35 ml
- Material:Zirconium oxide
- Seq:16
- Max. feed size:10 mm
- Sample volume:15 ml
Complementary Products