Order Entry
Japan
Orders LinkContactUsLinkComponent
 

VWR®, LED蛍光キューブ

UV LED fluorescence cube for IT406/407, LED emission 365 nm,
Excitation filter 325 - 375 nm,
Dichroic mirror cut-off 415 nm,
Emission filter 435 LP nm
  630-3260
VWR®, LED蛍光キューブ
  630-3260
VWR®, LED蛍光キューブ

UV LED fluorescence cube for IT406/407, LED emission 365 nm,
Excitation filter 325 - 375 nm,
Dichroic mirror cut-off 415 nm,
Emission filter 435 LP nm

 
 
   
1 * 1 items
 
  • 説明:
    UV LED fluorescence cube for IT406/407
  • Seq:
    04
  • Optical features:
    LED emission 365 nm,
    Excitation filter 325 - 375 nm,
    Dichroic mirror cut-off 415 nm,
    Emission filter 435 LP nm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.