VWR®, 安全洗浄ボトル
500 ml, Methylene dichloride, Natural bottle, wide neck, yellow cap
Catalog # 215-4305
Supplier: VWR Collection
Jump to:
Product Details & Documents
Integrated safety vent cap eliminates dripping of vapours.
- Caution note for each chemical
- Coloured caps
- Ergonomic spray angle
Bottles are manufactured from virgin LDPE resin, and come supplied with a polypropylene cap moulded with an ergonomic spray angle. The coloured caps allow for easy identification. A fill line is provided for user reference.
Please read the caution note mentioned over each bottle as per guideline.
注意: LDPE wash bottles are intended for short term dispensing use only and not for storage.
Specifications
- Type:Natural bottle, wide neck, yellow cap
- 容量:500 ml
- Seq:12
- Imprint:Methylene dichloride