Orders LinkContactUsLinkComponent
VWR®, 安全洗浄ボトル
250 ml, Methanol, Natural bottle, wide neck, green cap
Catalog # 215-4299
Supplier:   VWR Collection
VWR®, 安全洗浄ボトル
Catalog # 215-4299
Supplier:  VWR Collection
VWR®, 安全洗浄ボトル
VWR®, 安全洗浄ボトル
250 ml, Methanol, Natural bottle, wide neck, green cap
Price
Inquire For Price
Log in for availability
1 * 6 items
Product Details & Documents
Integrated safety vent cap eliminates dripping of vapours.
  • Caution note for each chemical
  • Coloured caps
  • Ergonomic spray angle
Bottles are manufactured from virgin LDPE resin, and come supplied with a polypropylene cap moulded with an ergonomic spray angle. The coloured caps allow for easy identification. A fill line is provided for user reference.
Please read the caution note mentioned over each bottle as per guideline.
 : LDPE wash bottles are intended for short term dispensing use only and not for storage.
Specifications
  • Type:
    Natural bottle, wide neck, green cap
  • 容量:
    250 ml
  • Seq:
    6
  • Imprint:
    Methanol