Order Entry
Japan
Orders LinkContactUsLinkComponent
 

VWR®ピペットチップ

Standard, 10 µl, +, Grey/red, 96 tips/rack, 10 racks/box, 2 boxes/case
Catalog # 612-6745
Supplier:   VWR Collection
VWR®ピペットチップ
Catalog # 612-6745
Supplier:  VWR Collection
VWR®ピペットチップ
VWR®ピペットチップ

Standard, 10 µl, +, Grey/red, 96 tips/rack, 10 racks/box, 2 boxes/case

Price
$55.97
Log in for availability
1 * 1.920 items
Product Details & Documents

These VWR® pipette tips are designed for precise liquid transfer.

  • Fully autoclavable to 121 °C
  • Colour-coded racks for easy volume identification
  • Tips with PP filter are also available
  • DNase-/RNase-free and non-pyrogenic
  • Every rack or case is printed with a batch number for traceability
  • Sterile tips are sterilized by irradiation SAL 10⁻⁶ (ISO 11137)
  • Shelf Life: 3 years after month of production

Good quality consumable products for micropipettors. Preferred accessories for most brand micropipettors.

 : Manufactured in a class 100 000 room environment under ISO 13485 and ISO 9001 quality management system.

Specifications
  • Colour code:
    Grey/red
  • Packed:
    96 tips/rack, 10 racks/box, 2 boxes/case
  • Sterile:
    +
  • 容量:
    10 µl
  • Seq:
    04
  • Tip style:
    Standard