Japan
ContactUsLinkComponent
2.2 ml square deep V bottom 96 well plate 26 cases of 50 plates packed on a Euro pallet Designed for compatibility with the Thermo Scientific Kingfisher Clean room manufactured as Dnase/RNAse free - Sarstedt - 1 * 1.300 items
Catalog # 733-2852
Supplier:  VWR Collection
2.2 ml square deep V bottom 96 well plate 26 cases of 50 plates packed on a Euro pallet Designed for compatibility with the Thermo Scientific Kingfisher Clean room manufactured as Dnase/RNAse free - Sarstedt - 1 * 1.300 items
Catalog # 733-2852
Supplier:  VWR Collection
Frequently Bought Together