Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, 試験管、フチなし

VWR®, 試験管、フチなし

5 ml, 75 mm, 12 mm, PS, Non graduated Sorwall type
  212-0729
VWR®, 試験管、フチなし
  212-0729
VWR®, 試験管、フチなし
VWR®, 試験管、フチなし

5 ml, 75 mm, 12 mm, PS, Non graduated Sorwall type

 
 
   
1 * 2.000 items
 

Polystyrene (PS), transparent or polypropylene (PP), translucent.

Universal tubes with round bottom for a wide range of applications.

 
  • 外径:
    12 mm
  • Type:
    Non graduated Sorwall type
  • 容量:
    5 ml
  • 長さ:
    75 mm
  • Material:
    PS
  • Seq:
    06
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.