VWR®, 試験管、赤色キャップ付き、無菌
5 ml, 75 mm, 13 mm, PS, Bulk
Supplier: VWR Collection
Jump to:
Product Details & Documents
Polystyrene (PS), clear or polypropylene (PP), natural.
Universal tubes with round bottom for a wide range of applications.
Documents
Certificates
Search by Lot NumberCoC Avantor Test Tubes with Red Cap, Sterile CoC VWR Test tubes with red cap, sterile.pdf CoC VWR® Test tubes with red cap, sterile CoC_VWRI212-0726_20F1335 CoC_VWRI212-0726_21F1258
More
CoC Avantor VWR® Test Tubes with Red Cap, Sterile.pdf
Conformity declaration
VWR® for sampling and sample transportation
Product Literature
Specifications
- 外径:13 mm
- Packed:Bulk
- 容量:5 ml
- 長さ:75 mm
- Material:PS
- Seq:03