Orders LinkContactUsLinkComponent
安全キャップ、VWR
安全キャップ、VWR
Safety cap with filter valve, GL 45, 3 port and 1 filter
Catalog # 590-0059
Supplier:   VWR Collection
安全キャップ、VWR
Catalog # 590-0059
Supplier:  VWR Collection
安全キャップ、VWR
安全キャップ、VWR
Safety cap with filter valve, GL 45, 3 port and 1 filter
Price
$90.25
Log in for availability
1 * 4 items
Product Details & Documents
VWR® safety cap extracts solvents or mobile phases safely from the bottle or reservoirs without letting the hazardous vapour come out. It filters particulates in the air and prevents contaminants and dusts entering solvents.
  • Advanced vapour valve keeps lab environment free from hazardous solvent vapour
  • Efficient PTFE filter for preventing contaminants mediated noise
  • Specially designed O-ring for air tight connection with glass bottle
  • Freely rotating cap body prevents twisting tubes during solvent bottle exchange
Safety Caps are recommended to use with HPLC solvents bottles for safer lab environment and lesser uncertainty of analysis.
Available with different number of ports. E.g. 1, 2, 3, 4 and 5.
Ordering information: One box contains set of four caps and their accessories.
A new box will have 4 sets of port blocks (with hole and without hole) in it.
Caution: The valve is recommended to be replaced after six months of use.
Documents
Specifications
  • Thread:
    GL 45
  • 説明:
    Safety cap with filter valve
  • Seq:
    03
  • Connector:
    3 port and 1 filter
Accessories
Product Information
  • Catalog #
    説明
    Availability
    Price
  • 590-0062
    Safety cap with filter valve accessories
    Log in for availability
    1 * 4 items - $40.70