Orders LinkContactUsLinkComponent
全酸価(TAN)標準
TAN005, 0,49 mg KOH/g, ASTM D664/IP 177, Glass bottle, 1×125 g, 125 g
Catalog # 99344.202
Supplier:   VWR Chemicals
全酸価(TAN)標準
Catalog # 99344.202
Supplier:  VWR Chemicals
CAS Number:  
全酸価(TAN)標準
全酸価(TAN)標準
TAN005, 0,49 mg KOH/g, ASTM D664/IP 177, Glass bottle, 1×125 g, 125 g
CAS Number:  
Price
$1,019.65
Log in for availability
1 * 125 g
Product Details & Documents
Total Acid Number (TAN) standards are specifically manufactured for the verification of analytical instruments used to determine acid number by potentiometric titration. Each standard is certified in strict accordance with international test method protocols, ASTM D664 / IP 177.
  • Typically used in, but not limited to, the analysis of used oils and lubricants
  • Acid number (AN) tested and certified in accordance with ASTM D664 / IP 177
  • Fully traceable to international standards and low levels of uncertainty achieves maximum accuracy of data
  • Supplied in tamper-evident glass packaging, providing assurance of sample integrity
 : Manufacture and certification is carried out in strict accordance with dual-UKAS accreditations to ISO 17025 and ISO 17034. This dual-accreditation ensures the highest level of accreditation guarantee, and provides the most credible certified data available worldwide.
 : The concentration value given in the ordering table below is the nominal concentration for the TAN value - actual batch values are given in the certification documents supplied with the product and also on the label.
Specifications
Specifications
  • 濃度:
    0,49 mg KOH/g
  • パックタイプ:
    Glass bottle
  • Pk:
    125 g
  • 説明:
    TAN005
  • Pk Info:
    1×125 g
  • Test method:
    ASTM D664/IP 177