Orders LinkContactUsLinkComponent
VWR® PHPピペットスタンド
PHP Shelf mounting stand for Pipette
  VWRA613-6724
VWR® PHPピペットスタンド
  VWRA613-6724
VWR® PHPピペットスタンド
VWR® PHPピペットスタンド
PHP Shelf mounting stand for Pipette
1 * 1 items
The VWR® Pipette Stand is a rotatable stand that conveniently holds up to six single and multi-channel pipettes. This helps users to maintain not only a tidy lab environment, but it also reduces the risk of pipette damage or cross-contamination.
  • Rotating (carousel) stand structure
  • Can hold up to six pipettes
  • Robust design
  • Efficient use of space
  • 説明:
    PHP Shelf mounting stand for Pipette