VWR® PHPピペットスタンド
PHP Shelf mounting stand for Pipette
VWRA613-6724
The VWR® Pipette Stand is a rotatable stand that conveniently holds up to six single and multi-channel pipettes. This helps users to maintain not only a tidy lab environment, but it also reduces the risk of pipette damage or cross-contamination.
- Rotating (carousel) stand structure
- Can hold up to six pipettes
- Robust design
- Efficient use of space
- 説明:PHP Shelf mounting stand for Pipette