Orders LinkContactUsLinkComponent
VWR® PHPピペットスタンド
VWR® PHPピペットスタンド
PHP Carousel stand for 6 pipettes
Catalog # VWRA613-6723
Supplier:   VWR Collection
VWR® PHPピペットスタンド
Catalog # VWRA613-6723
Supplier:  VWR Collection
VWR® PHPピペットスタンド
VWR® PHPピペットスタンド
PHP Carousel stand for 6 pipettes
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents
The VWR® Pipette Stand is a rotatable stand that conveniently holds up to six single and multi-channel pipettes. This helps users to maintain not only a tidy lab environment, but it also reduces the risk of pipette damage or cross-contamination.
  • Rotating (carousel) stand structure
  • Can hold up to six pipettes
  • Robust design
  • Efficient use of space
Specifications
  • 説明:
    PHP Carousel stand for 6 pipettes