Orders LinkContactUsLinkComponent
VWR® PHPピペット
2 - 20 µl
Catalog # VWRA613-6718
Supplier:   VWR Collection
VWR® PHPピペット
Catalog # VWRA613-6718
Supplier:  VWR Collection
VWR® PHPピペット
VWR® PHPピペット
2 - 20 µl
Product Details & Documents
VWR® PHP Pipettes are fully autoclavable, high performance pipettes with low plunger force.
  • Superior accuracy & precision (tested in ISO 17025 accredited lab)
  • Magnet assisted piston for low plunger force and lesser chance of RSI
  • PVDF tip cone for durable performance
  • Large and clear 4 digit display; color coding for easy identification
  • 3-Year warranty
VWR® PHP Pipettes are Precise High Performance, fully autoclavable pipettes, that are available in Single and Multi Channel range. VWR® PHP offers one of the lowest plunger forces and a durable tip cone. The tip ejector is made using special corrosion resistant alloy, protecting it against chemical and physical corrosion. The pipette has an effortless tip ejection with unique shock absorbing mechanism. The tip cone is spring loaded for lesser strain and proper tip fitment for precise pipetting (for Multi Channel only).
Specifications
  • 容量:
    2 - 20 µl
  • Seq:
    31