Orders LinkContactUsLinkComponent
電子ピペット、クラシック、VWR®。
電子ピペット、クラシック、VWR®。
10 - 200 µl
Catalog # VWRA613-6673
Supplier:   VWR Collection
電子ピペット、クラシック、VWR®。
Catalog # VWRA613-6673
Supplier:  VWR Collection
電子ピペット、クラシック、VWR®。
電子ピペット、クラシック、VWR®。
10 - 200 µl
Product Details & Documents
Precise 3 point calibrated electronic pipettes with 5 different liquid handling modes.
  • Auto-sensing magnetic piston for precise pipetting; State-of-the-art stepper motor for long lasting performance
  • Lightweight construction makes pipetting effortless and pain-free even after extensive usage
  • 5 different pipetting modes for all pipetting needs in a l ab: Forward pipetting, Reverse pipetting, Multi dispensing, Mixing, Diluting
  • Calibrated as per ISO 8655-6 standards in ISO 17025 accredited lab; 3 point auto-calibration provides most accurate and precise delivery of liquid throughout the volume range
  • Ultra-efficient battery allows 5 hours of continuous usage after every full recharge; Intelligent sleep mode conserves battery when not in use
  • A smart charging stand which can be used for charging or to rest the pipette when not in use
Intuitive graphical interface, wide display, multiple pipetting modes and ergonomic design redefines pipetting experience.
Certifications: CE, ISO8655 certified, tested in ISO17025 accredited lab.
Specifications
  • 容量:
    10 - 200 µl
  • Seq:
    03