シーリングメンブレン、クラシック、VWR®
Aluminum membrane
Catalog # VWRA734-3332
Supplier: VWR Collection
Jump to:
Product Details & Documents
VWR® PCR membranes are DNAse- and RNAse-free.
Minimises sample loss due to vaporisation
- Eliminates chances of leaching
Preferred solution for sealing.
Specifications
- 説明:Aluminum membrane
- Seq:03