シーリングメンブレン、クラシック、VWR®
PCR 96-well plate sealing membrane
Catalog # VWRA734-3330
Supplier: VWR Collection
Jump to:
Product Details & Documents
VWR® PCR membranes are DNAse- and RNAse-free.
Minimises sample loss due to vaporisation
- Eliminates chances of leaching
Preferred solution for sealing.
Documents
Avantor Liquid Handing and Sample Preparation Consumables
Product Literature
Specifications
- 説明:PCR 96-well plate sealing membrane
- Seq:01