VWR®, 臨床用4ウェイチューブラック、
Clinical 4-way tube racks, Natural, 12/18/12/18, 13/17 mm
Catalog # 525-1488
Supplier: VWR Collection
Jump to:
Product Details & Documents
These tube racks have a six way connection to create various configurations and are designed to hold round, flat and conical blood collection and culture tubes.
- Uniformly spaced wells help eliminate pipetting errors which enables more accurate results
- Molded alphanumeric identification for quick location and retrieval of tubes
- Made from polypropylene that is chemical resistant and easy to clean
- Racks can be used in water baths and placed in freezer
- Accommodates all brands of 13 and 17 mm tubes
They can be used as standalone unit or infinite units can be linked to create configurations to fit number of wells and tube sizes needed.
Specifications
- For tubes:13/17 mm
- 穴の数:12/18/12/18
- Colour:Natural
- 説明:Clinical 4-way tube racks