VWR®, アクセサリー
Peltier unit, A-1200; controller, constant-temperature system A1200, used for UV1600PC, UV-3100PC, V3000PC, UV6300PC, cell no. 1, path length: 10 mm, temp. range: 10 ~ 90 °C [0,1 °C interval adjustable (temp. control accuracy : ±0,1 °C)]
Catalog # 634-1154
Supplier: VWR Collection
Jump to:
Product Details & Documents
Specifications
- 説明:Peltier unit, A-1200; controller, constant-temperature system A1200, used for UV1600PC, UV-3100PC, V3000PC, UV6300PC, cell no. 1, path length: 10 mm, temp. range: 10 ~ 90 °C [0,1 °C interval adjustable (temp. control accuracy : ±0,1 °C)]
- Seq:03