Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, クライオボックスおよび仕切り

VWR®, クライオボックスおよび仕切り

16, 125×125×25,4 mm
Catalog # 479-2225
Supplier:   VWR Collection
VWR®, クライオボックスおよび仕切り
Catalog # 479-2225
Supplier:  VWR Collection
VWR®, クライオボックスおよび仕切り
VWR®, クライオボックスおよび仕切り

16, 125×125×25,4 mm

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents

Cardboard, white.

Boxes and dividers are constructed from durable fibreboard with a protective, moisture-repellant coating.

Boxes with dividers feature a lid printed with a numbered grid for superior inventory control. Boxes without dividers can accommodate dividers (sold separately) for 16 to 100 places. Fibreboard dividers can fit in any 127×127 mm cryogenic or mechanical freezer box.

Specifications
  • Compartments:
    16
  • W×D×H ext.:
    125×125×25,4 mm
  • Seq:
    07
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.