Orders LinkContactUsLinkComponent
VWR®, クライオボックスおよび仕切り
25, 133x133x 25,4 mm
  479-2224
VWR®, クライオボックスおよび仕切り
  479-2224
VWR®, クライオボックスおよび仕切り
VWR®, クライオボックスおよび仕切り
25, 133x133x 25,4 mm
1 * 1 items
Cardboard, white.
Boxes and dividers are constructed from durable fibreboard with a protective, moisture-repellant coating.
Boxes with dividers feature a lid printed with a numbered grid for superior inventory control. Boxes without dividers can accommodate dividers (sold separately) for 16 to 100 places. Fibreboard dividers can fit in any 127×127 mm cryogenic or mechanical freezer box.
  • Compartments:
    25
  • W×D×H ext.:
    133x133x 25,4 mm
  • Seq:
    14