Order Entry
Japan
Orders LinkContactUsLinkComponent
VWR®, クライオボックスおよび仕切り

VWR®, クライオボックスおよび仕切り

49, 125×125×25,4 mm
Catalog # 479-2223
Supplier:   VWR Collection
VWR®, クライオボックスおよび仕切り
Catalog # 479-2223
Supplier:  VWR Collection
VWR®, クライオボックスおよび仕切り
VWR®, クライオボックスおよび仕切り

49, 125×125×25,4 mm

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents

Cardboard, white.

Boxes and dividers are constructed from durable fibreboard with a protective, moisture-repellant coating.

Boxes with dividers feature a lid printed with a numbered grid for superior inventory control. Boxes without dividers can accommodate dividers (sold separately) for 16 to 100 places. Fibreboard dividers can fit in any 127×127 mm cryogenic or mechanical freezer box.

Specifications
  • Compartments:
    49
  • W×D×H ext.:
    125×125×25,4 mm
  • Seq:
    15