VWR®, クライオボックスおよび仕切り
81, 125×125×25,4 mm
479-2221
Cardboard, white.
Boxes and dividers are constructed from durable fibreboard with a protective, moisture-repellant coating.
Boxes with dividers feature a lid printed with a numbered grid for superior inventory control. Boxes without dividers can accommodate dividers (sold separately) for 16 to 100 places. Fibreboard dividers can fit in any 127×127 mm cryogenic or mechanical freezer box.
- Compartments:81
- W×D×H ext.:125×125×25,4 mm
- Seq:23