Order Entry
Japan
Orders LinkContactUsLinkComponent
 

VWR®, クライオボックスおよび仕切り

Cryobox without divider, waterproof, White, 127×127×73,0 mm
Catalog # 479-2220
Supplier:   VWR Collection
VWR®, クライオボックスおよび仕切り
Catalog # 479-2220
Supplier:  VWR Collection
VWR®, クライオボックスおよび仕切り
VWR®, クライオボックスおよび仕切り

Cryobox without divider, waterproof, White, 127×127×73,0 mm

Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents

Cardboard, white.

Boxes and dividers are constructed from durable fibreboard with a protective, moisture-repellant coating.

Boxes with dividers feature a lid printed with a numbered grid for superior inventory control. Boxes without dividers can accommodate dividers (sold separately) for 16 to 100 places. Fibreboard dividers can fit in any 127×127 mm cryogenic or mechanical freezer box.

Specifications
  • W×D×H ext.:
    127×127×73,0 mm
  • Colour:
    White
  • 説明:
    Cryobox without divider, waterproof
  • Seq:
    01