Orders LinkContactUsLinkComponent
L字型セルスプレッダー、無菌
L字型セルスプレッダー、無菌
Green, Individually packed
Catalog # VWRU76208-438
Supplier:   VWR Collection
L字型セルスプレッダー、無菌
Catalog # VWRU76208-438
Supplier:  VWR Collection
L字型セルスプレッダー、無菌
L字型セルスプレッダー、無菌
Green, Individually packed
Price
$10.46
Log in for availability
1 * 100 items
Product Details & Documents
Disposable L-shaped cell spreaders are ideal for spreading cell or bacterial cultures evenly.
  • L-shape allows application on the entire dish or plate surface
  • Smooth spreading surface with slight upward return reduces the risk of damage to samples
  • Sterilised by electron beam irradiation
  • Non-pyrogenic
Spreaders made of polypropylene and pre-sterilised, eliminating the need for flaming or autoclaving.
Certifications: Manufactured in a class 100000 cleanroom environment; ISO13485:2003 and ISO 9001:2008.
Packaging: Individual wrap and bulk packaging available.
Specifications
  • Packed:
    Individually packed
  • Colour:
    Green
  • Seq:
    01