Order Entry
Japan
Orders LinkContactUsLinkComponent
 

VWR®, 吸引ピペット

10 ml, 343 mm, Individually wrapped, 50/bag
  612-5886
VWR®, 吸引ピペット
  612-5886
VWR®, 吸引ピペット
VWR®, 吸引ピペット

10 ml, 343 mm, Individually wrapped, 50/bag

 
 
   
1 * 200 items
 

PS pipettes, non plugged, non graduated, disposable, sterile.

  • Use at –20 to +50 °C
  • Sterilised by gamma irradiation SAL 10⁻⁶ (ISO 11137)
  • Shelf life of 4 years after month of production
  • Free of detectable DNase and RNase
  • Non pyrogenic, non cytotoxic

For aseptically aspirating liquid by using vacuum suction.

 : Products are manufactured under the ISO 13485 standard. Meet the requirements of BSE/TSE.

 
  • Packed:
    Individually wrapped, 50/bag
  • 容量:
    10 ml
  • 長さ:
    343 mm
  • Seq:
    04