Orders LinkContactUsLinkComponent
L字型セルスプレッダー、無菌
Green, Bulk packed; 10/bag
  VWRU76207-748
L字型セルスプレッダー、無菌
  VWRU76207-748
L字型セルスプレッダー、無菌
L字型セルスプレッダー、無菌
Green, Bulk packed; 10/bag
$28.83
1 * 500 items
Disposable L-shaped cell spreaders are ideal for spreading cell or bacterial cultures evenly.
  • L-shape allows application on the entire dish or plate surface
  • Smooth spreading surface with slight upward return reduces the risk of damage to samples
  • Sterilised by electron beam irradiation
  • Non-pyrogenic
Spreaders made of polypropylene and pre-sterilised, eliminating the need for flaming or autoclaving.
 : Manufactured in a class 100000 cleanroom environment; ISO13485:2003 and ISO 9001:2008.
 : Individual wrap and bulk packaging available.
  • Packed:
    Bulk packed; 10/bag
  • Colour:
    Green
  • Seq:
    02