L字型セルスプレッダー、無菌
Green, Bulk packed; 10/bag
VWRU76207-748
Disposable L-shaped cell spreaders are ideal for spreading cell or bacterial cultures evenly.
- L-shape allows application on the entire dish or plate surface
- Smooth spreading surface with slight upward return reduces the risk of damage to samples
- Sterilised by electron beam irradiation
- Non-pyrogenic
Spreaders made of polypropylene and pre-sterilised, eliminating the need for flaming or autoclaving.
: Manufactured in a class 100000 cleanroom environment; ISO13485:2003 and ISO 9001:2008.
: Individual wrap and bulk packaging available.
- Packed:Bulk packed; 10/bag
- Colour:Green
- Seq:02