Orders LinkContactUsLinkComponent
VWR®, バケツライナー、滅菌
VWR®, バケツライナー、滅菌
Bucket liners for B-7 buckets, sterile, LDPE/nylon, 790×480 mm
Catalog # 141-1443
Supplier:   VWR Collection
VWR®, バケツライナー、滅菌
Catalog # 141-1443
Supplier:  VWR Collection
VWR®, バケツライナー、滅菌
VWR®, バケツライナー、滅菌
Bucket liners for B-7 buckets, sterile, LDPE/nylon, 790×480 mm
Price
Inquire For Price
Log in for availability
1 * 72 items
Product Details & Documents
Bucket liners create an impermeable barrier between the disinfectant and receptacle.
  • LDPE/nylon blend
  • Irradiated
  • Fits B-7 series buckets
Helps ensure the integrity of a disinfectant blend.
包装: Individually bagged, double-bagged in groups of 6. 4 packs of 6 per case.
Specifications
  • 説明:
    Bucket liners for B-7 buckets, sterile
  • 長さ×幅:
    790×480 mm
  • Material:
    LDPE/nylon
  • Seq:
    02