VWR®, スプラッシュヘッド、Stutzer、ストレート
29/32 NS, 29/32 NS
Supplier: VWR Collection
Jump to:
Product Details & Documents
Borosilicate 3.3 glass.
- Very good chemical resistance
- High temperature resistance and high resistance to thermal shock
- Straight, with standard ground joint and socket
These splash heads, according to Stutzer, can be placed directly onto the distillation flask to intercept droplets.
Specifications
- Cone size:29/32 NS
- Socket size:29/32 NS