VWR®, スプラッシュヘッド、Stutzer、ストレート
19/26 NS, 24/29 NS
Catalog # 201-1400
Supplier: VWR Collection
Jump to:
Product Details & Documents
Borosilicate 3.3 glass.
- Very good chemical resistance
- High temperature resistance and high resistance to thermal shock
- Straight, with standard ground joint and socket
These splash heads, according to Stutzer, can be placed directly onto the distillation flask to intercept droplets.
Specifications
- Cone size:24/29 NS
- Socket size:19/26 NS