VWR®, スプラッシュヘッド、Stutzer、ストレート
14/23 NS, 14/23 NS
201-1398
Borosilicate 3.3 glass.
- Very good chemical resistance
- High temperature resistance and high resistance to thermal shock
- Straight, with standard ground joint and socket
These splash heads, according to Stutzer, can be placed directly onto the distillation flask to intercept droplets.
- Cone size:14/23 NS
- Socket size:14/23 NS