VWR® MicroStar 12, ローター
Rotor for 10×1,8 ml cryotubes, max. RCF 12300 ×g
Catalog # 521-2650
Supplier: VWR Collection
Jump to:
Product Details & Documents
Documents
521-1652
Images 360
Centrifugation Solutions from Avantor
Product Literature
Specifications
- 説明:Rotor for 10×1,8 ml cryotubes, max. RCF 12300 ×g
- Seq:012