Order Entry
Japan
Orders LinkContactUsLinkComponent
 

クロマトグラフィーチューブ, PTFE

1,58 mm, 3,18 mm, 5 m
  08-PTFE-62-5M
 :   HICHROM
クロマトグラフィーチューブ, PTFE
  08-PTFE-62-5M
 :  HICHROM
クロマトグラフィーチューブ, PTFE
クロマトグラフィーチューブ, PTFE

1,58 mm, 3,18 mm, 5 m

 
 
   
1 * 1 items
 

PTFE

  • Flexible and economical
  • Excellent solvent resistance

Inert to all chemicals used in HPLC but suitable for low-pressure applications only. The most common tubing used to connect the solvent reservoir to the pump.

 
  • 外径:
    3,18 mm
  • 内径:
    1,58 mm
  • 長さ:
    5 m
  • Seq:
    15
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.