Order Entry
Japan
Orders LinkContactUsLinkComponent
 

コンタクトスライド、VWR

Contact slides for total bacterial count (agar type: PCA + TTC + disinfectant neutralisers) and enterobacteria (agar type: VRBG + disinfectant neutralisers), 20
  525262V
コンタクトスライド、VWR
  525262V
 :  
コンタクトスライド、VWR
コンタクトスライド、VWR

Contact slides for total bacterial count (agar type: PCA + TTC + disinfectant neutralisers) and enterobacteria (agar type: VRBG + disinfectant neutralisers), 20

 :  
 
$124.67
   
1 * 20 items
 

Surface and liquid flexible slides for simple and effective microbiological monitoring and hygiene testing of surfaces and fluids.

  • Two tests with one slide: Each slide is coated on both sides either with the same agar or with a different agar on each side
  • Long shelf life: 9 months from production
  • Clear instructions allow a simple way to evaluate the microbiological contamination of fluid surfaces (low, medium or high)
  • Specifically formulated for total bacterial count and for detecting yeast and moulds, enterobacteria, E. coli and coliform
  • Easily used and transported away from the laboratory to control the surfaces of production plants
  • Easy to use thanks to the screw cap
 

 

  • Pk:
    20
  • 説明:
    Contact slides for total bacterial count (agar type: PCA + TTC + disinfectant neutralisers) and enterobacteria (agar type: VRBG + disinfectant neutralisers)

 

Accessories

 

  •  
    Type
    Shelves supplied (max.)
     
     
  • 390-0384
    INCU-Line® IL 10 with transparent window
    1 (2)
       
    1 * 1 items

 

  •  
    Type
    Shelves supplied (max.)
     
     
  • 390-0385
    INCU-Line® IL 10 with tinted Plexiglas® window
    1 (2)
       
    1 * 1 items

 

  •  
    説明
     
     
  • 390-0386
    Tube rack for dip slides, 2×6 positions, Ø 32 mm
       
    1 * 1 items