Order Entry
Japan
Orders LinkContactUsLinkComponent
コンタクトスライド、VWR

コンタクトスライド、VWR

Contact slides for total bacterial count (agar type: PCA + TTC + disinfectant neutralisers) and for the detection of E. coli and coliform (agar type: chromogenic and selective medium that allows the detection of E. coli and differentiation from other enterobacteria). Note Minimum Order Quantity is 50 pieces, 20
Catalog # 525392V
Supplier:   VWR Chemicals
コンタクトスライド、VWR
Catalog # 525392V
Supplier:  VWR Chemicals
CAS Number:  
コンタクトスライド、VWR
コンタクトスライド、VWR

Contact slides for total bacterial count (agar type: PCA + TTC + disinfectant neutralisers) and for the detection of E. coli and coliform (agar type: chromogenic and selective medium that allows the detection of E. coli and differentiation from other enterobacteria). Note Minimum Order Quantity is 50 pieces, 20

CAS Number:  
Price
$132.72
Log in for availability
1 * 20 items
Product Details & Documents

Surface and liquid flexible slides for simple and effective microbiological monitoring and hygiene testing of surfaces and fluids.

  • Two tests with one slide: Each slide is coated on both sides either with the same agar or with a different agar on each side
  • Long shelf life: 9 months from production
  • Clear instructions allow a simple way to evaluate the microbiological contamination of fluid surfaces (low, medium or high)
  • Specifically formulated for total bacterial count and for detecting yeast and moulds, enterobacteria, E. coli and coliform
  • Easily used and transported away from the laboratory to control the surfaces of production plants
  • Easy to use thanks to the screw cap
Specifications

Specifications

  • Pk:
    20
  • 説明:
    Contact slides for total bacterial count (agar type: PCA + TTC + disinfectant neutralisers) and for the detection of <i>E. coli</i> and coliform (agar type: chromogenic and selective medium that allows the detection of <i>E. coli</i> and differentiation from other enterobacteria). Note Minimum Order Quantity is 50 pieces

Specifications

Accessories

Product Information

  • Catalog #
    Type
    Shelves supplied (max.)
    Availability
    Price
  • 390-0384
    INCU-Line® IL 10 with transparent window
    1 (2)
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    Type
    Shelves supplied (max.)
    Availability
    Price
  • 390-0385
    INCU-Line® IL 10 with tinted Plexiglas® window
    1 (2)
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 390-0386
    Tube rack for dip slides, 2×6 positions, Ø 32 mm
    Log in for availability
    1 * 1 items