Order Entry
Japan
Orders LinkContactUsLinkComponent
コンタクトスライド、VWR

コンタクトスライド、VWR

Contact slides for total bacterial count (agar type: TSA+ disinfectant neutralisers) on both sides of the slide, 20
Catalog # 525472V
Supplier:   VWR Chemicals
コンタクトスライド、VWR
Catalog # 525472V
Supplier:  VWR Chemicals
CAS Number:  
コンタクトスライド、VWR
コンタクトスライド、VWR

Contact slides for total bacterial count (agar type: TSA+ disinfectant neutralisers) on both sides of the slide, 20

CAS Number:  
Price
$138.99
Log in for availability
1 * 20 items
Product Details & Documents

Surface and liquid flexible slides for simple and effective microbiological monitoring and hygiene testing of surfaces and fluids.

  • Two tests with one slide: Each slide is coated on both sides either with the same agar or with a different agar on each side
  • Long shelf life: 9 months from production
  • Clear instructions allow a simple way to evaluate the microbiological contamination of fluid surfaces (low, medium or high)
  • Specifically formulated for total bacterial count and for detecting yeast and moulds, enterobacteria, E. coli and coliform
  • Easily used and transported away from the laboratory to control the surfaces of production plants
  • Easy to use thanks to the screw cap
Specifications

Specifications

  • Pk:
    20
  • 説明:
    Contact slides for total bacterial count (agar type: TSA+ disinfectant neutralisers) on both sides of the slide

Specifications

Accessories

Product Information

  • Catalog #
    Type
    Shelves supplied (max.)
    Availability
    Price
  • 390-0384
    INCU-Line® IL 10 with transparent window
    1 (2)
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    Type
    Shelves supplied (max.)
    Availability
    Price
  • 390-0385
    INCU-Line® IL 10 with tinted Plexiglas® window
    1 (2)
    Log in for availability
    1 * 1 items

Product Information

  • Catalog #
    説明
    Availability
    Price
  • 390-0386
    Tube rack for dip slides, 2×6 positions, Ø 32 mm
    Log in for availability
    1 * 1 items

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.