UV/Visibleスタンダード
UV/Vis standard 5, toluene solution in n-hexane for resolution testing, 1 KIT
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
For spectrophotometer calibration according to European Pharmacopoeia (Ph. Eur.)
- Absorbance control
- Testing of resolution power
- Stray light testing
- Wavelength accuracy testing
In the pharmaceutical industry, a regular check of UV/Visible spectrophotometers according to Ph. Eur. is mandatory. Even in other industries, researchers who are using UV/Visible spectrophotometers, and who are working according to e.g. GMP, GLP, USP or ISO guidelines, need to check and qualify their instruments on a regular basis.
Specifications
Specifications
- Pk:1 KIT
- 説明:UV/Vis standard 5, toluene solution in n-hexane for resolution testing