Orders LinkContactUsLinkComponent
UV/Visibleスタンダード
UV/Visibleスタンダード
UV/Vis standard 3, for stray light testing, 1 KIT
Catalog # 86127.001
Supplier:   VWR Chemicals
UV/Visibleスタンダード
Catalog # 86127.001
Supplier:  VWR Chemicals
CAS Number:  
UV/Visibleスタンダード
UV/Visibleスタンダード
UV/Vis standard 3, for stray light testing, 1 KIT
CAS Number:  
Price
$371.09
Log in for availability
1 * 1 KIT
Product Details & Documents
For spectrophotometer calibration according to European Pharmacopoeia (Ph. Eur.)
  • Absorbance control
  • Testing of resolution power
  • Stray light testing
  • Wavelength accuracy testing
In the pharmaceutical industry, a regular check of UV/Visible spectrophotometers according to Ph. Eur. is mandatory. Even in other industries, researchers who are using UV/Visible spectrophotometers, and who are working according to e.g. GMP, GLP, USP or ISO guidelines, need to check and qualify their instruments on a regular basis.
Documents
Certificates
Search by Lot Number
Specifications
Specifications
  • Pk:
    1 KIT
  • 説明:
    UV/Vis standard 3, for stray light testing
Specifications