Order Entry
Japan
Orders LinkContactUsLinkComponent
 

Propionic acid ≥99% (by GC), J.T.Baker®

Glass bottle, amber, narrow mouth, 4 L
  U330-09
 
 
 
 :   Avantor
Propionic acid ≥99% (by GC), J.T.Baker®
  U330-09
 :  Avantor
 :  79-09-4
Propionic acid ≥99% (by GC), J.T.Baker®
Propionic acid ≥99% (by GC), J.T.Baker®

Glass bottle, amber, narrow mouth, 4 L

 :  79-09-4
 
 
   
1 * 4 L
 
 :  Propanoic acid
  •  :79-09-4
  •  :C₃H₆O₂
  •  :MFCD00002756
  •  :74.08 g/mol
  •  :13,07917
  •  :141 °C (1013 hPa)
  •  :–21 °C
  •  :0,99 g/cm³ (20 °C)
  •  :49 °C
  •  :Ambient
  •  :201-176-3
  •  :3463
  •  :8 (3),II
  •  :01-2119486971-24

 

 

 

  • Assay (CH₃CH₂COOH) (by GC):
    ≥99 %

 

  • パックタイプ:
    Glass bottle, amber, narrow mouth
  • Pk:
    4 L